missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes

Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.

Brand:  Eppendorf™ 0030123344

Code : 96

Additional Details : Weight : 1.21500kg

 View more versions of this product

Product Code. 10073321

Quantity Price
1 £69.75 / Pack of 1000
10 £64.55 / Pack of 1000
In stock, ready to ship: 22

Add to basket


Description

Description

Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.

  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Guaranteed functionality from -86 to 100°C
  • Large frosted lid and surface on the side for easy labeling
  • Autoclavable when open (121°C, 20 min)
Specifications

Specifications

Autoclavable
Polypropylene
Natural
1000
Yes
Centrifuge Tube
2 mL
For all applications in molecular biology, clinical chemistry and biochemistry
Videos
SDS
Documents
Certificates