missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCNA Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding mature form of human PCNA (Met 1-Ser 261) fused to polyhistidine tag at N-terminus

Brand:  Thermo Scientific™ 12131H07B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15295775

  • £206.12 / 25µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

  • Formulation: 50mM Na3PO4, 300mM NaCl, 10% Glycerol, 2mM DTT, pH 7.0
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5111
≥85% by SDS-PAGE
Human
<1 EU/μg
PCNA
Other Proteins
PCNA Recombinant Human Protein
Lyophilized
31kd
5 x 50μg
Full Length, Recombinant
Proliferating Cell Nuclear Antigen
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.