missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PCNA Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding mature form of human PCNA (Met 1-Ser 261) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 12131H07B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15225785

  • £308.14 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

  • Formulation: 50mM Na3PO4, 300mM NaCl, 10% Glycerol, 2mM DTT, pH 7.0
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5111
≥85% by SDS-PAGE
Human
<1 EU/μg
PCNA
Other Proteins
PCNA Recombinant Human Protein
Lyophilized
31kd
50μg
Full Length, Recombinant
Proliferating Cell Nuclear Antigen
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.