missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 1X), sterile-filtered

 

Brand:  Thermo Scientific Chemicals J61196.AP

25.26 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.00050kg

Product Code. 15453819

  • £27.10 / 500mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phosphate-buffered saline (PBS, 1X), sterile is a specifically formulated buffer solution and is used for reconstituting sterile cytokines or antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

500 mL
1 X
Phosphate Buffered Saline (PBS)
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only