Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), with Triton|r X-100
Ideal for maintaining a constant pH.
£41.60 - £69.60
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15410317
|
Thermo Scientific Chemicals
J63521.AP |
500 mL |
£69.60
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15400317
|
Thermo Scientific Chemicals
J63521.AK |
250 mL |
£41.60
250mL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.