missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
£55.35 - £163.00
Specifications
Content And Storage | Room temperature |
---|---|
Packaging Type | Poly Cube |
Melting Point | 0°C |
pH | 7.3 to 7.5 |
Form | Liquid |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
10091403
|
Fisher Bioreagents
BP2438-20 |
20L | Poly Cube |
£163.00
20 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10173433
|
Fisher Bioreagents
BP2438-4 |
4L | Poly Cube |
£55.35
4 L |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
A standard phosphate buffer used in many biomolecular procedures.Specifications
Room temperature | |
0°C | |
Liquid |
Poly Cube | |
7.3 to 7.5 |