Learn More
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
Brand: Fisher Bioreagents BP2438-20
Code : 32
Additional Details : CAS Number : 7558-79-4 Weight : 20.05000kg
Descripción
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X PBS buffer solution
- Eliminates the hassle of dilution from stock solutions or weighing and mixing individual components
- 1X solution contains 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
- Filtered and tested for the absence of DNase, RNase, and protease
- PBS is used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Especificaciones
Room temperature | |
0°C | |
7.3 to 7.5 |
Poly Cube | |
20L | |
Liquid |