Learn More
IBA Lifesciences CD81 Isolation Kit for FABian™
IBA Lifesciences™ CD81 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
Brand:  IBA Lifesciences 6-6015-001
Additional Details : Weight : 0.17000kg
 View more versions of this product
Product Code. 15809784
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The kit is used for the selection of PBMCs from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD81 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 9mL whole blood.
Specifications
Specifications
Isolation of label-free cells
2°C to 8°C
9 mL Whole Blood
Cell Isolation Kit
Human Whole Blood
1 Kit, Human