missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551033
Additional Details : Weight : 0.01000kg
Description
PAX1 Polyclonal Antibody detects endogenous levels of PAX1 protein.
PAX1 Polyclonal antibody specifically detects PAX1 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
PAX1 | |
Polyclonal | |
Unconjugated | |
PAX1 | |
hbs; hunchback; HuP48; OFC2; paired box 1; paired box gene 1; Paired box homeotic gene 1; paired box protein Pax-1; paired domain gene HuP48; PAX1; Pax-1; un; undulated; wavy tail; wt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5075 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P15863 | |
PAX1 | |
Synthesized peptide derived from the Internal region of human Pax-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |