missing translation for 'onlineSavingsMsg'
Learn More

Patulin, 99%, Thermo Scientific Chemicals

Catalog Number p-178335
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Packaging:
Glass bottle
149-29-1
C7H6O4
154.12
MFCD00005858
ZRWPUFFVAOMMNM-UHFFFAOYSA-N
patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin
4696
CHEBI:74926
4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
C1C=C2C(=CC(=O)O2)C(O1)O
149-29-1
C7H6O4
154.12
MFCD00005858
ZRWPUFFVAOMMNM-UHFFFAOYSA-N
patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin
4696
CHEBI:74926
4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
C1C=C2C(=CC(=O)O2)C(O1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 108.0°C to 111.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 7158
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethyl and amyl acetate,insoluble in petroleum ether
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Patulin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Patulin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'