missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PASK Recombinant Human Protein
PASK is an evolutionarily conserved protein present in yeast, flies, and mammals.
£1604.00 - £6900.00
Specifications
Accession Number | NP_055963 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 77.2kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448503
|
Invitrogen™
PR7013A |
100μg |
£1604.00
100µg |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10270283
|
Invitrogen™
PV4276 |
1mg |
£6900.00
1mg |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PASK domains regulate the function of many intracellular signaling pathways in response to both extrinsic and intrinsic stimuli.
Specifications
NP_055963 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
77.2kDa | |
RUO - research use only | |
PASK | |
Human | |
GST Tag | |
Frozen |