Learn More
3M™ Particulate Respirator, K100 Series
Provides comfortable, solid, and professional protection at an affordable price.
Brand:  3M™ 7000108735
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11755993
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Provides comfortable, solid, and professional protection at an affordable price.
Specifications
Specifications
EN149:2001+A1 2009 NR D
Diamond-shaped front, single loop strap
Yes
Exposure to dust particles and /or solid and non-volatile liquid particles
Diamond shape front
White
FFP1
FFP1
Includes yellow coloured noseclip for easy identification (FFP1)