Learn More
PARP1 Monoclonal Antibody (HC2R8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£86.25 - £211.00
Specifications
Antigen | PARP1 |
---|---|
Clone | HC2R8 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15277927
|
Affymetrix eBioscience
14-6667-82 |
100 μg |
£211.00
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15267927
|
Affymetrix eBioscience
14-6667-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This HC2R8 monoclonal antibody reacts with human poly (ADP-ribose) polymerase (PARP1). This ubiquitous 116 kDa nuclear enzyme is involved in DNA repair. During apoptosis, active caspases -3, -6 and -7 cleave PARP1 after Asp214, thereby inactivating PARP1 and generating two apoptotic fragments sized 85 kDa and 25 kDa. The HC2R8 antibody specifically recognizes both the cleaved and non-cleaved forms of PARP. Applications Reported: This HC2R8 antibody has been reported for use in immunoblotting (WB). Applications Tested: This HC2R8 antibody has been tested by immunoblotting of staurosporine-treated Jurkat cells. This can be used at less than or equal to 0.1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Specifications
PARP1 | |
0.5 mg/mL | |
Unconjugated | |
Mouse | |
Human | |
142 | |
Primary | |
4° C |
HC2R8 | |
Monoclonal | |
Liquid | |
RUO | |
P09874 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.