Learn More
Cole-Parmer™ Parker Hannifin™ Flare Compression Pipe Adapter Elbow
Leak-tight and high-pressure connection for corrosive chemical applications
Brand: Cole-Parmer™ 30490-74
Code : NEW
Additional Details : Weight : 0.04000kg
Description
- PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance
- Leaf-free design withstands pressures to 225psi
- Ideal for high-vibration or side-load stressed applications
- Low dead volume reduces particle entrapment, limits bacterial growth in deionized water and simplifies rinsing
Specifications
Flare Compression Pipe Adapter | |
PFA | |
1/2 in. | |
Translucent |
Flare x NPT(M) Connection | |
Corrosive Chemical Applications | |
1/2 in. NPT(M) |