missing translation for 'onlineSavingsMsg'
Learn More

Parker domnick hunter™ Ultra-High Purity Hydrogen Generators for GC and GC/MS

Ultra-high purity, low maintenance generators supply hydrogen for GC and GC/MS carrier gas applications, plus all commonly used combustion detectors used routinely in GC

Brand:  Parker domnick hunter™ 40H-MD

Code : 04

Additional Details : Weight : 20.50000kg

 View more versions of this product

Product Code. 12862636

  • £10915.02 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Generate hydrogen from just deionized water and electricity, at low pressure and only on demand. Choose from three diffferent models, with flowrates ranging from 160mL/min. to 500mL/min. All models feature unique generator monitoring and alarm system that ensures high reliability and user safety.

  • Ion exchange membrane produces a flow of ultra-pure hydrogen; regenerative desiccant dryer purifies it further for use as a carrier gas
  • Gas on demand: no warmup period
  • Cell compression and water management units ensure cell reliability and longevity
  • Monitoring/alarm system provides automatic hydrogen leak detection and continuous monitoring of vital parameters
  • Seven-stage explosion protection system plus minimal stored gas mean safe operation

Products in this group have a universal power supply and can be used with any voltage 100-230V 50/60Hz.

Compliance

CE, CSA and UL compliant and listed.

Instrument & Equipment Services

Two years, cell; one year, entire system

Specifications

Specifications

Hydrogen Generator
47cm
45.6cm
5° to 40°C
90 to 264V
90 to 264V 50/60Hz
99.9999%
210w
CE, CSA, UL
250mL/min.
50/60Hz
0.125 in. compression fitting
25kg
40H-MD
10 to 100psi
Videos