missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline base, Thermo Scientific Chemicals

Catalog Number p-178309
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
467-62-9
C19H19N3O
305.38
MFCD00036222
KRVRUAYUNOQMOV-UHFFFAOYSA-N
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
10084
tris(4-aminophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N
This item is not returnable. View return policy
467-62-9
C19H19N3O
305.38
MFCD00036222
KRVRUAYUNOQMOV-UHFFFAOYSA-N
pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
10084
tris(4-aminophenyl)methanol
C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 205.0°C
Color Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 13, 733
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 305.38
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline base
compliance-icons
Product Identifier
  • Pararosaniline base
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'