missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraffin, liquid, technical, Thermo Scientific Chemicals
128.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 L
Packaging:
Plastic Can
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 8012-95-1 |
Melting Point | -24.0°C |
Density | 0.8800g/mL |
Boiling Point | 300.0°C |
Flash Point | 218°C |
Packaging | Plastic Can |
Molecular Formula | MFCD00131611 |
MDL Number | MFCD00131611 |
Quantity | 25 L |
Specific Gravity | 0.88 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'