missing translation for 'onlineSavingsMsg'
Learn More

Pancuronium dibromide, Tocris Bioscience™

Nicotinic (neuromuscular) antagonist

Brand:  Tocris (Bio-Techne) 069350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487953

  • £266.00 / 50mg


Estimated Shipment
08-03-2018
Add to basket

Description

Description

Nicotine (neuromuscular) antagonist. Skeletal muscle relaxant.
Specifications

Chemical Identifiers

Pancuronium dibromide
C35H60Br2N2O4
732.68
1,1'-(3α,17β-Dihydroxy-2β,5α-androstan-2β,16β-ylene) bis[1-methylpiperidinium] diacetate dibromide

Specifications

Pancuronium dibromide
>99%
732.68
50mg
15500-66-0
C35H60Br2N2O4
1,1'-(3α,17β-Dihydroxy-2β,5α-androstan-2β,16β-ylene) bis[1-methylpiperidinium] diacetate dibromide
Videos