missing translation for 'onlineSavingsMsg'
Learn More

PAN, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391239
Click to view available options
Quantity:
1 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
85-85-8
C15H11N3O
249.273
RAXUMGMWXZYADR-OBGWFSINSA-N
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
5376264
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3
This item is not returnable. View return policy
85-85-8
C15H11N3O
249.273
RAXUMGMWXZYADR-OBGWFSINSA-N
1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
5376264
(1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Quantity 1 g
Formula Weight 249.28
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material PAN
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'