Learn More
Palmitoylisopropylamide, Tocris Bioscience™
Inhibitor of FAAH
Brand:  Tocris (Bio-Techne) 181550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13430064
  • £176.40 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Inhibitor of fatty acid amide hydrolase (FAAH); pIC50 = 4.89 for inhibition of [3H]-anandamide metabolism. Displays little binding to CB1 and CB2 receptors (IC50 > 100 μM) and very weakly blocks anandamide uptake (IC50 ∽ 100 μM). Inhibits proliferation of C6 glioma cells.
Specifications
Chemical Identifiers
189939-61-5
297.527
palmitoylisopropylamide, chembl32827, hexadecanoic acid isopropylamide, n-isopropylhexadecanamide, palmitoyl-n-isopropylamide, n-1-methylethyl-hexadecanamide, n-isopropylpalmitamide, tocris-1815, n-isopropyl-hexadecanamide, palmitoyl n-isopropylamide
N-propan-2-ylhexadecanamide
C19H39NO
TZYVUGCYYQOTQR-UHFFFAOYSA-N
5063962
CCCCCCCCCCCCCCCC(=O)NC(C)C
Specifications
Palmitoylisopropylamide
189939-61-5
palmitoylisopropylamide, chembl32827, hexadecanoic acid isopropylamide, n-isopropylhexadecanamide, palmitoyl-n-isopropylamide, n-1-methylethyl-hexadecanamide, n-isopropylpalmitamide, tocris-1815, n-isopropyl-hexadecanamide, palmitoyl n-isopropylamide
CCCCCCCCCCCCCCCC(=O)NC(C)C
297.527
297.52
50mg
C19H39NO
TZYVUGCYYQOTQR-UHFFFAOYSA-N
N-propan-2-ylhexadecanamide
5063962
>98%