missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium wire, 0.406mm (0.016 in.) dia., as drawn, 99.95% (metals basis)

Brand:  Alfa Aesar™ 045123.BY

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.01000kg

Quantity 100cm

Product Code. 15496755

  • £183.27 / 100cm


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Specifications

Palladium wire
100cm
7440-05-3
99.95%
MFCD00011167
≈1.56g/m
0.406mm (0.016 in.) dia., as drawn
Wire
(metals basis)
Odorless
Videos
SDS