missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium Thinfoil, 0.005mm (0.0002 in.) thick, 99.95% (metals basis)

Brand:  Alfa Aesar™ 042690.FF

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.01000kg

Quantity 25x25mm

 View more versions of this product

Product Code. 11305437

  • £435.02 / 25mm


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium Thinfoil
7440-05-3
0.005mm (0.0002 in.) thick, Light tight, no visible pinholes
MFCD00011167

Specifications

Palladium Thinfoil
25x25mm
7440-05-3
99.95%
MFCD00011167
≈0.037g/25x25mm
0.005mm (0.0002 in.) thick, Light tight, no visible pinholes
Thin Foil
(metals basis)
Odorless
Videos
SDS