Learn More
Thermo Scientific Alfa Aesar Palladium Thinfoil, 0.005mm (0.0002 in.) thick, 99.95% (metals basis)
Brand: Thermo Scientific Alfa Aesar 042690.FI
Palladium Thinfoil, 0.005mm (0.0002 in.) thick, 99.95% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-05-3 Weight : 0.01000kg