missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium Silver foil, 0.127mm (0.005 in.) thick, 99.9% (metals basis excluding Pt)

Brand:  Alfa Aesar™ 012575.GH

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100x100mm

 View more versions of this product

Product Code. 11307427

  • £934.70 / Each


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium Silver foil
0.127mm (0.005 in.) thick
MFCD02091738

Specifications

Pd:Ag; 75:25 wt%
≈0.92g/25x25mm
0.127mm (0.005 in.) thick
99.9%
MFCD02091738
Palladium Silver foil
100x100mm
Foil
(metals basis excluding Pt)
Odorless
Videos
SDS