missing translation for 'onlineSavingsMsg'
Learn More

Palladium Powder, Alfa Aesar™

Brand:  Alfa Aesar 011721.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11341808

  • £28.50 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Palladium
5% on barium carbonate powder
Powder
MFCD03427451

Specifications

Palladium
5% on barium carbonate powder
gray powder
MFCD03427451
Gray
5g
7440-05-3
Powder
Odorless
Videos
SDS