missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium Molybdenum wire, 0.5mm (0.0197 in.) dia.

Brand:  Alfa Aesar™ 044245.BY

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100cm

 View more versions of this product

Product Code. 11319778

  • £411.28 / 100cm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Palladium Molybdenum wire
0.5mm (0.0197 in.) dia.
MFCD03792509

Specifications

Pd:Mo:Pt; 85.5:13.5:1 approx. wt%
≈1.17g/50cm
0.5mm (0.0197 in.) dia.
Wire
1700°C
Palladium Molybdenum wire
100cm
11.9g/cm3
MFCD03792509
Videos
SDS