missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-3737138
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
42196-31-6
C4F6O4Pd
332.45
MFCD00013204
PBDBXAQKXCXZCJ-UHFFFAOYSA-L
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
2733347
palladium(2+);2,2,2-trifluoroacetate
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
42196-31-6
C4F6O4Pd
332.45
MFCD00013204
PBDBXAQKXCXZCJ-UHFFFAOYSA-L
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
2733347
palladium(2+);2,2,2-trifluoroacetate
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 210°C
Color Blue to Blue-Green
Infrared Spectrum Authentic
Assay Percent Range 30.9 to 33.2% (Pd)
Packaging Glass bottle
Linear Formula (CF3CO2)2Pd
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 332.44
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Palladium(II) trifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'