missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium(II) sulfate dihydrate, Premion™, 99.95% (metals basis), Pd 44.1% min

Brand:  Alfa Aesar™ 012165.03

Code : NEW

Additional Details : CAS Number : 13444-98-9 Weight : 0.00100kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11396538

  • £63.20 / 1g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Palladium(II) sulfate dihydrate
Premion™
238.51 (202.48 Anhydrous)
UN3260
13444-98-9
PdSO4·2H2O
MFCD00011173

Specifications

Palladium(II) sulfate dihydrate
Premion™
99.95%
PdSO4·2H2O
MFCD00011173
Pd 44.1% min.
Red to brown
13444-98-9
Crystalline
(metals basis)
238.51 (202.48 Anhydrous)
UN3260
Hygroscopic
1g
Videos
SDS