missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) oxide, 86.8-87.1% Pd, ACROS Organics™

Brand:  Acros Organics 195160050

Code : 29

Additional Details : CAS Number : 1314-08-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10296630

  • £276.50 / 5g


Estimated Shipment
15-02-2018
Add to basket

Specifications

Chemical Identifiers

Palladium(II) oxide
1314-08-5
OPd
122.4
86.8-87.1% Pd
86.8 to 87.1% (Pd)
PdO
MFCD00011172

Specifications

Palladium(II) oxide
1314-08-5
86.8 to 87.1% (Pd)
PdO
MFCD00011172
Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Glass bottle
Gray to black
5g
86.8-87.1% Pd
Powder
OPd
122.4
15, 7094
8.7
Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
870.0°C
Videos
SDS