missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium(II) nitrate, solution, Pd 4-5% w/w (cont. Pd)

Brand:  Alfa Aesar™ 045512.NB

Code : NEW

Additional Details : CAS Number : 10102-05-3 Weight : 0.00500kg Transport : UN number : 3264 Chem class : 1a Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 15416985

  • £190.40 / 5g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Palladium (II) nitrate solution
N2O6Pd
230.43
UN3264
10102-05-3
Pd(NO3)2
MFCD00011169

Specifications

Palladium (II) nitrate solution
Liquid
Pd 4 to 5% w/w (cont. Pd)
230.43
UN3264
5g
10102-05-3
N2O6Pd
Pd(NO3)2
MFCD00011169
Brown to yellow to orange
Videos
SDS