missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium(II) cyanide, Pd 66%

Brand:  Alfa Aesar™ 011879.03

Code : NEW

Additional Details : CAS Number : 2035-66-7 Weight : 0.00100kg Transport : UN number : 1588 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11391958

  • £205.64 / 1g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium(II) cyanide
158.74
UN1588
2035-66-7
MFCD00016282

Specifications

Palladium(II) cyanide
Powder
MFCD00016282
Pd 66%
2035-66-7
158.74
UN1588
1g
Videos
SDS