missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water

Brand:  Alfa Aesar™ 045576.09

Code : NEW

Additional Details : CAS Number : 12135-22-7 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15454955

  • £80.60 / 10g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium hydroxide
12135-22-7
Pd 5% on carbon powder, Type A403002-5, Nominally 50% water
MFCD00064599

Specifications

Palladium hydroxide
12135-22-7
MFCD00064599
10g
Pd 5% on carbon powder, Type A403002-5, Nominally 50% water
Powder
Black
Odorless
Videos
SDS