missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on charcoal paste, Type 487

Brand:  Alfa Aesar™ H36938.14

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15467717

  • £97.80 / 25g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on charcoal paste, Type 487
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Dark gray
5% on charcoal paste, Type 487
Solid
Odorless
25g
Videos
SDS