missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on carbon, Type 87L, nominally 50% water wet

Brand:  Alfa Aesar™ H33434.09

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15438465

  • £133.90 / 10g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
5% on carbon, Type 87L, nominally 50% water wet

Specifications

Palladium
7440-05-3
Dark gray
5% on carbon, Type 87L, nominally 50% water wet
Powder
10g
Videos
SDS