missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on carbon paste, A405028-5

Brand:  Alfa Aesar™ H36996.14

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15447727

  • £135.90 / 25g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on carbon paste, A405028-5
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Dark gray
5% on carbon paste, A405028-5
Solid
Odorless
25g
Videos
SDS