missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 5% on calcium carbonate, lead poisoned, A305060-5, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Gray |
Physical Form | Powder |
Quantity | 5 g |
Odor | Odorless |
Chemical Name or Material | Palladium, 5% on calcium carbonate, lead poisoned, A305060-5 |
Safety and Handling
Product Identifier
- Palladium, 5% on calcium carbonate, lead poisoned,A305060-5
- Danger
- Reproductive toxicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 2
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Lead
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'