missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on calcium carbonate, lead poisoned, A305060-5

Brand:  Alfa Aesar™ H36186.22

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15409075

  • £620.80 / 100g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
5% on calcium carbonate, lead poisoned, A305060-5
Lindlar catalyst
7440-05-3
MFCD03427452

Specifications

Palladium
7440-05-3
MFCD03427452
Odorless
100g
5% on calcium carbonate, lead poisoned, A305060-5
Powder
Lindlar catalyst
Dark gray
Videos
SDS