missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on calcium carbonate, A303060-5

Brand:  Alfa Aesar™ H36622.14

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15427677

  • £67.90 / 25g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on calcium carbonate, A303060-5
MFCD03427452

Specifications

Palladium
7440-05-3
MFCD03427452
Dark brown or gray to black
5% on calcium carbonate, A303060-5
Powder
Odorless
25g
Videos
SDS