missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet

Brand:  Alfa Aesar™ 044142.30

Code : NEW

Additional Details : Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 11359638

  • £903.40 / 250g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Description

Description

  • Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
Specifications

Chemical Identifiers

Palladium
5% on activated carbon powder, Standard, Reduced, Acidic catalyst, Nominally 50% water wet
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Odorless
5% on activated carbon powder, Standard, Reduced, Acidic catalyst, Nominally 50% water wet
Powder
250g
900 to 1100m2/g
Videos
SDS