missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on activated carbon paste, Type 452

Brand:  Alfa Aesar™ H37443.22

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15447777

  • £228.30 / 100g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on activated carbon paste, Type 452
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Dark gray
5% on activated carbon paste, Type 452
Solid
Odorless
100g
Videos
SDS