missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 3% on activated carbon powder, eggshell, reduced

Brand:  Alfa Aesar™ 011691.14

Code : NEW

Additional Details : Weight : 0.02500kg Transport : UN number : 3178 Chem class : 4.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11341798

  • £59.20 / 25g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
3% on activated carbon powder, Eggshell, Reduced
MFCD03457879
UN3178

Specifications

Palladium
7440-05-3
UN3178
Odorless
3% on activated carbon powder, Eggshell, Reduced
MFCD03457879
25g
900 to 1100m2/g
Videos
SDS