missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 20% on activated carbon powder, eggshell, reduced, nominally 50% water wet

Brand:  Alfa Aesar™ 038306.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11364417

  • £1330.40 / 100g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
20% on activated carbon powder, Eggshell, reduced, nominally 50% water wet
MFCD03457879

Specifications

Palladium
7440-05-3
100g
20% on activated carbon powder, Eggshell, reduced, nominally 50% water wet
MFCD03457879
900 to 1100m2/g
Videos
SDS