missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 10% on carbon, Type 487, nominally 50% water wet

Brand:  Alfa Aesar™ H34200.18

Code : NEW

Additional Details : Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 15458825

  • £301.70 / 50g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
10% on carbon, Type 487, nominally 50% water wet

Specifications

Palladium
7440-05-3
Dark gray
10% on carbon, Type 487, nominally 50% water wet
Powder
50g
Videos
SDS