missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 10% on carbon, Type 487, dry, Thermo Scientific Chemicals

Catalog Number 11438700
Click to view available options
Quantity:
2 g
10 g
50 g
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

Solubility
Insoluble in water and organic solvents.

Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
TRUSTED_SUSTAINABILITY
Quantity 2 g
UN Number UN1325
Merck Index 14,6989
Solubility Information Insoluble in water and organic solvents.
Chemical Name or Material Palladium, 10% on carbon, Type 487, Dry
compliance-icons
Product Identifier
  • Palladium, 10% on carbon, Type 487, dry
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : carbon, palladium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'