Alfa Aesar™ Palladium, 10% on carbon paste, Type 755


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H37277.22

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 7440-05-3 Weight: 0.10000kg


Quantity 100g

Product Code. 15487747

Quantity Price
1 £ 307.19 / 100g
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 10% on carbon paste, Type 755
MDL Number MFCD03457879

Specification

Color Dark gray
Odor Odorless
Name Note 10% on carbon paste, Type 755
Chemical Name or Material Palladium
MDL Number MFCD03457879
CAS 7440-05-3
Physical Form Solid
TSCA TSCA
Quantity 100g
Documentation

Technical documents