missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet

Brand:  Alfa Aesar™ 046789.04

Code : NEW

Additional Details : Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15444085

  • £21.60 / 2g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
10% on activated carbon powder, Type A402023-10, Nominally 50% water wet
MFCD03457879

Specifications

Palladium
Sold On A Dry Weight Basis
Black
10% on activated carbon powder, Type A402023-10, Nominally 50% water wet
MFCD03457879
2g
Videos
SDS