missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar Palladium, 10% on activated carbon powder, Type 87L, standard, reduced, nominally 50% water wet
Brand: Alfa Aesar 045924.14
Palladium, 10% on activated carbon powder, Type 87L, standard, reduced, nominally 50% water wet
Code : NEW
Additional Details : Weight : 0.02500kg