Learn More
Alfa Aesar™ Palladium, 10% on activated carbon paste, Type 39
Brand: Alfa Aesar™ H37431.14
Code : NEW
Additional Details : CAS Number : 7440-05-3 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
Palladium | |
7440-05-3 | |
10% on activated carbon paste, Type 39 | |
MFCD03457879 |
Specifications
Palladium | |
7440-05-3 | |
MFCD03457879 | |
Dark gray |
10% on activated carbon paste, Type 39 | |
Solid | |
Odorless | |
25g |