missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 1% on granular carbon, reduced

Brand:  Alfa Aesar™ 046462.36

Code : NEW

Additional Details : Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15432697

  • £1123.40 / 500g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
1% on granular carbon, Reduced
MFCD00011167

Specifications

Palladium
Pellet
500g
1% on granular carbon, Reduced
MFCD00011167
3mm (0.13 in.)
Videos
SDS