missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Palladium, 1% on granular carbon, reduced
Brand: Alfa Aesar™ 046462.22
Code : NEW
Additional Details : Weight : 0.10000kg
Quantity | 100g |
---|
Chemical Identifiers
Palladium | |
1% on granular carbon, Reduced | |
MFCD00011167 |
Specifications
Palladium | |
Pellet | |
100g |
1% on granular carbon, Reduced | |
MFCD00011167 | |
3mm (0.13 in.) |